3-Chlorocinnamic acid 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
3-(3-Chlorophenyl)acrylic acid, (2E)-3-(3-Chlorophenyl)acrylic acid
A254893-5G
A254893-100G
A254893-10G
A254893-500G
A254893-25G
103852-028EA
5
USD
103852-028
77091-486
77262-092
77262-094
77262-096
77304-642
3-Chlorocinnamic acid 98%
3-Chlorocinnamic acid
Precaución: for Research Use Only
Formula:
C₉H₇ClO₂ MW: 182.61 g/mol Boiling Pt: 326.2 °C (760 mm Hg) Melting Pt: 161…164 °C Flash Pt: 151.1 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004384 CAS Number: 1866-38-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...