3-Chlorobenzoylacetonitrile 97%

Supplier: Ambeed
Warning

Synonyms: 3'-Chloro-2-cyanoacetophenone, (3-Chlorobenzoyl)acetonitrile, 3-(3-Chlorophenyl)-3-oxopropanenitrile

A157962-10G A157962-1G A157962-5G A157962-25G
103805-072EA 20 USD
103805-072 103843-278 77259-496 77300-306 77319-120 77748-070
3-Chlorobenzoylacetonitrile 97%
3-Chlorobenzoylacetonitrile

Precaución: for Research Use Only

Formula: C₉H₆ClNO
MW: 179.61 g/mol
Boiling Pt: 347.1 °C (760 mm Hg)
Melting Pt: 78 °C
Flash Pt: 163.7 °C
Storage Temperature: Ambient
MDL Number: MFCD00067891
CAS Number: 21667-62-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR