3-(3-Bromophenyl)propionic acid 97%

Supplier: Ambeed
Warning

A160393-10G A160393-1G A160393-25G A160393-5G
103805-372EA 5 USD
103805-372 103843-600 77259-596 77300-460 77319-532 77319-534
3-(3-Bromophenyl)propionic acid 97%
3-(3-Bromophenyl)propionic acid

Precaución: for Research Use Only

Formula: C₉H₉BrO₂
Boiling Pt: 336.3 °C (760 mm Hg)
Melting Pt: 76 °C
Flash Pt: 157.2 °C
Storage Temperature: Ambient
MDL Number: MFCD01310792
CAS Number: 42287-90-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR