3-(3,5-Di-tert-butyl-4-hydroxyphenyl)propionic acid 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
3,5-Di-tert-butyl-4-hydroxyphenylpropionic acid
A229376-25G
A229376-100G
A229376-10G
A229376-500G
A229376-1000G
103850-372EA
9
USD
103850-372
77089-214
77550-480
77550-482
77600-838
77667-908
3-(3,5-Di-tert-butyl-4-hydroxyphenyl)propionic acid 97%
3-(3,5-Di-tert-butyl-4-hydroxyphenyl)propionic acid
Precaución: for Research Use Only
Formula:
C₁₇H₂₆O₃ MW: 278.40 g/mol Boiling Pt: 364.2 °C (760 mm Hg) Melting Pt: 172…174 °C Flash Pt: 188.3 °C Storage Temperature: Ambient |
MDL Number:
MFCD00017519 CAS Number: 20170-32-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...