3-(3,5-Dimethoxyphenyl)propionic acid 98%

Supplier: Ambeed
Warning

A150921-100G A150921-10G A150921-250MG A150921-1G A150921-25G A150921-5G
103804-216EA 6 USD
103804-216 103842-356 77137-766 77137-768 77137-770 77299-838 77317-890 77317-892
3-(3,5-Dimethoxyphenyl)propionic acid 98%
3-(3,5-Dimethoxyphenyl)propionic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₄O₄
MW: 210.23 g/mol
Boiling Pt: 363.8 °C (760 mm Hg)
Melting Pt: 60…62 °C
Flash Pt: 141.1 °C
Storage Temperature: Ambient
MDL Number: MFCD03425685
CAS Number: 717-94-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR