3,5-Dimethoxycinnamic acid 98%

Supplier: Ambeed
Warning

Synonyms: 3-(3,5-Dimethoxyphenyl)acrylic acid

A872872-1G A872872-5G A872872-10G
103841-288EA 32 USD
103841-288 76845-544 76845-546 77281-816
3,5-Dimethoxycinnamic acid 98%
3,5-Dimethoxycinnamic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₂O₄
MW: 208.21 g/mol
Boiling Pt: 377.1±27.0 °C (760 mm Hg)
Melting Pt: 174…175 °C
Flash Pt: 148.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00004389
CAS Number: 16909-11-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR