3-(3,4-Dimethoxyphenyl)propionic acid 98%

Supplier: Ambeed
Warning

Synonyms: 3,4-Dimethoxyhydrocinnamic acid, 3-(3,4-Dimethoxyphenyl)propanoic acid

A268016-100G A268016-10G A268016-5G
103852-792EA 13 USD
103852-792 76772-100 76772-102 77304-994
3-(3,4-Dimethoxyphenyl)propionic acid 98%
3-(3,4-Dimethoxyphenyl)propionic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₄O₄
MW: 210.23 g/mol
Boiling Pt: 344.2 °C (760 mm Hg)
Melting Pt: 96…97 °C
Flash Pt: 132 °C
Storage Temperature: Ambient
MDL Number: MFCD00002774
CAS Number: 2107-70-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR