3-[3-(Trifluoromethyl)phenyl]propanal 98%

Supplier: Ambeed
Warning

Synonyms: 3-(3-(Trifluoromethyl)phenyl)propanal, 3-[3-(Trifluoromethyl)phenyl]propaldehyde, 3-(3-Oxoprop-1-yl)benzotrifluoride, 3-[3-(Trifluoromethyl)phenyl]propionaldehyde

A285597-100MG A285597-1G A285597-5G
103813-682EA 42 USD
103813-682 103834-816 77032-218 77305-478 77305-480
3-[3-(Trifluoromethyl)phenyl]propanal 98%
3-[3-(Trifluoromethyl)phenyl]propanal

Precaución: for Research Use Only

Formula: C₁₀H₉F₃O
MW: 202.173 g/mol
Boiling Pt: 207.4 °C (760 mm Hg)
Flash Pt: 93.1 °C
Storage Temperature: Freezer
MDL Number: MFCD07772918
CAS Number: 21172-41-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR