3-(o-Tolyl)propionic acid 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
3-(2-Methylphenyl)propionic acid, 2-Methylhydrocinnamic acid, 3-(2-Methylphenyl)propanoic acid
A219211-100G
A219211-10G
A219211-250MG
A219211-1G
A219211-5G
103818-770EA
11
USD
103818-770
103849-644
76691-734
76691-736
76691-738
77088-190
77088-192
3-(o-Tolyl)propionic acid 98%
3-(o-Tolyl)propionic acid
Precaución: for Research Use Only
Formula:
C₁₀H₁₂O₂ MW: 164.2 g/mol Boiling Pt: 293.3 °C (760 mm Hg) Melting Pt: 100…103 °C Flash Pt: 190.5 °C Storage Temperature: Ambient |
MDL Number:
MFCD00079773 CAS Number: 22084-89-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...