3-(2-Methoxyphenyl)propanal ≥95%, Ambeed.Inc

Supplier: Ambeed
Warning

A258514-100MG A258514-250MG
103812-378EA 221 USD
103812-378 103852-256 77683-788 77890-456
3-(2-Methoxyphenyl)propanal ≥95%, Ambeed.Inc
3-(2-Methoxyphenyl)propanal

Precaución: For research use only.

Formula: C₁₀H₁₂O₂
Boiling Pt: 240.5 °C (760 mm Hg)
Flash Pt: 95.5 °C
Storage Temperature: Freezer
MDL Number: MFCD07772908
CAS Number: 33538-83-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR