3-(2-Methoxyphenyl)propan-1-ol 95%

Supplier: Ambeed
Warning

A184261-100MG A184261-250MG A184261-1G A184261-5G
103815-732EA 110 USD
103815-732 103846-294 77274-212 77274-214 77301-890 77301-892
3-(2-Methoxyphenyl)propan-1-ol 95%
3-(2-Methoxyphenyl)propan-1-ol

Precaución: for Research Use Only

Formula: C₁₀H₁₄O₂
Boiling Pt: 271.2 °C (760 mm Hg)
Flash Pt: 113.4 °C
Storage Temperature: Ambient
MDL Number: MFCD00197108
CAS Number: 10493-37-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR