3-(2-Hydroxyethyl)benzonitrile 97%

Supplier: Ambeed
Warning

A153415-100MG A153415-250MG A153415-1G A153415-5G
103804-512EA 302 USD
103804-512 103842-684 77235-526 77235-528 77318-340 77318-342
3-(2-Hydroxyethyl)benzonitrile 97%
3-(2-Hydroxyethyl)benzonitrile

Precaución: for Research Use Only

Formula: C₉H₉NO
Storage Temperature: Ambient
MDL Number: MFCD10001394
CAS Number: 193290-27-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR