3-(2-Fluorophenyl)propionic acid 97%

Supplier: Ambeed
Warning

A145561-10G A145561-25G A145561-1G
103803-554EA 8 USD
103803-554 103841-638 76689-158 77299-464 77316-944
3-(2-Fluorophenyl)propionic acid 97%
3-(2-Fluorophenyl)propionic acid

Precaución: for Research Use Only

Formula: C₉H₉FO₂
Boiling Pt: 273.7 °C (760 mm Hg)
Melting Pt: 76 °C
Flash Pt: 119.3 °C
Storage Temperature: Ambient
MDL Number: MFCD01310820
CAS Number: 1643-26-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR