3-(2-Chloropyrimidin-4-yl)-1-methyl-1H-indole 98%

Supplier: Ambeed
Warning

A104126-100G A104126-10G A104126-1G A104126-25G A104126-5G
103822-370EA 15 USD
103822-370 77160-362 77160-364 77160-366 77160-368 77309-232
3-(2-Chloropyrimidin-4-yl)-1-methyl-1H-indole 98%
3-(2-Chloropyrimidin-4-yl)-1-methyl-1H-indole

Precaución: for Research Use Only

Formula: C₁₃H₁₀ClN₃
MW: 243.69 g/mol
Boiling Pt: 486.3±28.0 °C (760 mm Hg)
Melting Pt: 200…204 °C
Flash Pt: 247.9 °C
Storage Temperature: Refrigerator
MDL Number: MFCD26397861
CAS Number: 1032452-86-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR