3-(2-Chlorophenyl)propionic acid 98%

Supplier: Ambeed
Warning

A285216-100G A285216-10G A285216-1G A285216-25G A285216-5G
103813-660EA 5 USD
103813-660 103834-796 76816-430 76816-432 76837-326 76837-328 76837-330
3-(2-Chlorophenyl)propionic acid 98%
3-(2-Chlorophenyl)propionic acid

Precaución: for Research Use Only

Formula: C₉H₉ClO₂
Boiling Pt: 306.6 °C (760 mm Hg)
Melting Pt: 94…96 °C
Flash Pt: 139.2 °C
Storage Temperature: Ambient
MDL Number: MFCD00016547
CAS Number: 1643-28-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR