3-(2-Chloroethyl)-2-methyl-6,7,8,9-tetrahydro-4H-pyrido[1,2-a]pyrimidin-4-one hydrochloride 98%
Supplier: Ambeed
|
Danger
|
A103258-250MG
A103258-1G
103796-520EA
12
USD
103796-520
103822-222
77270-334
77309-102
3-(2-Chloroethyl)-2-methyl-6,7,8,9-tetrahydro-4H-pyrido[1,2-a]pyrimidin-4-one hydrochloride 98%
3-(2-Chloroethyl)-2-methyl-6,7,8,9-tetrahydro-4H-pyrido[1,2-a]pyrimidin-4-one hydrochloride
Precaución: for Research Use Only
Formula:
C₁₁H₁₆Cl₂N₂O MW: 263.16 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09031252 CAS Number: 93076-03-0 UN: 3261 ADR: 8,Ⅱ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...