3-(2-Bromophenyl)propionic acid 98%
Supplier: Ambeed
|
Warning
|
A226095-25G
A226095-5G
A226095-100G
A226095-10G
A226095-500G
A226095-1G
103819-222EA
5
USD
103819-222
103850-124
77088-846
77088-848
77261-420
77261-422
77261-424
77303-760
3-(2-Bromophenyl)propionic acid 98%
3-(2-Bromophenyl)propionic acid
Precaución: for Research Use Only
Formula:
C₉H₉BrO₂ MW: 229.07 g/mol Boiling Pt: 331 °C (760 mm Hg) Melting Pt: 98…102 °C Flash Pt: 154 °C Storage Temperature: Ambient |
MDL Number:
MFCD01310791 CAS Number: 15115-58-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...