3-(2-Bromophenyl)-1-propanol 97%

Supplier: Ambeed
Warning

Synonyms: 3-(2-Bromo-phenyl)-propan-1-ol

A173186-10G A173186-250MG A173186-1G A173186-25G A173186-5G
103806-802EA 9 USD
103806-802 103845-096 76974-108 76974-110 77082-334 77082-336 77082-338
3-(2-Bromophenyl)-1-propanol 97%
3-(2-Bromophenyl)-1-propanol

Precaución: for Research Use Only

Formula: C₉H₁₁BrO
MW: 215.09 g/mol
Boiling Pt: 294 °C (760 mm Hg)
Flash Pt: 132.195 °C
Storage Temperature: Ambient
MDL Number: MFCD09028685
CAS Number: 52221-92-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR