3-(2,4-Dimethyl-1H-pyrrol-3-yl)propanoic acid 97%

Supplier: Ambeed
Warning

A315819-100MG A315819-250MG A315819-1G
103825-084EA 259 USD
103825-084 103836-006 77251-056 77251-058 77306-136
3-(2,4-Dimethyl-1H-pyrrol-3-yl)propanoic acid 97%
3-(2,4-Dimethyl-1H-pyrrol-3-yl)propanoic acid

Precaución: for Research Use Only

Formula: C₉H₁₃NO₂
MW: 167.21 g/mol
Boiling Pt: 332.8 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD02684431
CAS Number: 54474-50-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR