3-(2,4-Dihydroxyphenyl)propionic Acid 97%

Supplier: Ambeed
Warning

A100991-1MG A100991-250MG A100991-25MG A100991-50MG A100991-1G A100991-100MG
103795-738EA 65 USD
103795-738 103821-430 77270-190 77270-192 77270-194 77270-196 77296-936 77747-755
3-(2,4-Dihydroxyphenyl)propionic Acid 97%
3-(2,4-Dihydroxyphenyl)propionic Acid

Precaución: for Research Use Only

Formula: C₉H₁₀O₄
Boiling Pt: 418 °C (760 mm Hg)
Melting Pt: 158…162 °C
Storage Temperature: Ambient
MDL Number: MFCD00143017
CAS Number: 5631-68-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR