3-(2-(2-(2-Bromoethoxy)ethoxy)ethoxy)propanoic acid 97%

Supplier: Ambeed
Warning

Synonyms: Bromo-PEG3-C2-acid

A292036-100MG A292036-250MG A292036-5G
103799-578EA 68 USD
103799-578 103813-922 77277-178 77616-270 77748-570
3-(2-(2-(2-Bromoethoxy)ethoxy)ethoxy)propanoic acid 97%
3-(2-(2-(2-Bromoethoxy)ethoxy)ethoxy)propanoic acid

Precaución: for Research Use Only

Formula: C₉H₁₇BrO₅
MW: 285.13 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22574820
CAS Number: 782475-35-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR