3-Pyrrolidin-1-ylpropan-1-ol 95%

Supplier: Ambeed
Warning

Synonyms: 3-(1-Pyrrolidyl)-1-propanol

A264840-100MG A264840-10G A264840-250MG A264840-25G
103812-696EA 28 USD
103812-696 103852-590 77098-902 77098-904 77098-906 77304-906
3-Pyrrolidin-1-ylpropan-1-ol 95%
3-Pyrrolidin-1-ylpropan-1-ol

Precaución: for Research Use Only

Formula: C₇H₁₅NO
MW: 129.2 g/mol
Boiling Pt: 216.9 °C (760 mm Hg)
Flash Pt: 97.7 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01687769
CAS Number: 19748-66-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR