N-(2-Cyanoethyl)pyrrole 98%
Supplier: Ambeed
|
Warning
|
A337205-100G
A337205-10G
A337205-1G
A337205-25G
A337205-5G
103825-742EA
14
USD
103825-742
103836-716
76724-278
76724-280
76839-720
76839-722
76839-724
N-(2-Cyanoethyl)pyrrole 98%
N-(2-Cyanoethyl)pyrrole
Precaución: for Research Use Only
Formula:
C₇H₈N₂ Boiling Pt: 266.8 °C (760 mm Hg) Flash Pt: 113 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003092 CAS Number: 43036-06-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...