3-(1,3-Dioxolan-2-yl)propanoic acid 95%

Supplier: Ambeed
Warning

A224017-5G A224017-10G A224017-250MG A224017-1G
103819-076EA 74 USD
103819-076 103849-964 77088-594 77275-566 77275-568 77303-682
3-(1,3-Dioxolan-2-yl)propanoic acid 95%
3-(1,3-Dioxolan-2-yl)propanoic acid

Precaución: for Research Use Only

Formula: C₆H₁₀O₄
MW: 146.14 g/mol
Boiling Pt: 246.1 °C (760 mm Hg)
Melting Pt: 33…37 °C
Storage Temperature: Ambient
MDL Number: MFCD06209471
CAS Number: 4388-56-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR