3-(1,3-Dioxoisoindolin-2-yl)propanoic acid 98%

Supplier: Ambeed
Warning

Synonyms: 3-Phthalimidopropionic acid, 3-(1,3-dioxo-2,3-dihydro-1H-isoindol-2-yl)propanoic acid

A177033-25G A177033-5G A177033-100G A177033-10G
103845-528EA 6 USD
103845-528 77082-906 77082-908 77287-030 77287-032
3-(1,3-Dioxoisoindolin-2-yl)propanoic acid 98%
3-(1,3-Dioxoisoindolin-2-yl)propanoic acid

Precaución: for Research Use Only

Formula: C₁₁H₉NO₄
Boiling Pt: 423.4 °C (760 mm Hg)
Melting Pt: 151 °C
Storage Temperature: Ambient
MDL Number: MFCD00023096
CAS Number: 3339-73-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR