3-(1,3-Dioxo-1,3-dihydroisoindol-2-yl)propionaldehyde 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
3-(Phthalimid-1-yl)propanal, 3-(1,3-Dioxoisoindolin-2-yl)propanal
A201734-1G
A201734-5G
A201734-100MG
A201734-250MG
103817-322EA
47
USD
103817-322
103847-962
77086-040
77086-042
77274-864
77274-866
3-(1,3-Dioxo-1,3-dihydroisoindol-2-yl)propionaldehyde 95%
3-(1,3-Dioxo-1,3-dihydroisoindol-2-yl)propionaldehyde
Precaución: for Research Use Only
Formula:
C₁₁H₉NO₃ MW: 203.2 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD01704466 CAS Number: 2436-29-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...