3-(1-(tert-Butoxycarbonyl)piperidin-4-yl)propanoic acid 97%

Supplier: Ambeed
Warning

A194868-10G A194868-250MG A194868-1G A194868-25G A194868-5G A194868-100G
103816-700EA 45 USD
103816-700 103847-314 76721-256 76721-258 77085-248 77085-250 77085-252 77667-632
3-(1-(tert-Butoxycarbonyl)piperidin-4-yl)propanoic acid 97%
3-(1-(tert-Butoxycarbonyl)piperidin-4-yl)propanoic acid

Precaución: for Research Use Only

Formula: C₁₃H₂₃NO₄
MW: 257.3300 g/mol
Boiling Pt: 386.4 °C (760 mm Hg)
Melting Pt: 120…124 °C
Flash Pt: 187.5 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01861212
CAS Number: 154775-43-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR