3-((tert-Butoxycarbonyl)amino)cyclopentanecarboxylic acid 95%

Supplier: Ambeed
Warning

A132324-1G A132324-5G
103801-720EA 84 USD
103801-720 77314-298 77314-300
3-((tert-Butoxycarbonyl)amino)cyclopentanecarboxylic acid 95%
3-((tert-Butoxycarbonyl)amino)cyclopentanecarboxylic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₉NO₄
MW: 229.27 g/mol
Boiling Pt: 382.5±31.0 °C (760 mm Hg)
Flash Pt: 185.1 °C
Storage Temperature: Refrigerator
MDL Number: MFCD02093997
CAS Number: 855863-93-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR