Boc-DL-β-Aib-OH 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
3-((tert-Butoxycarbonyl)amino)-2-methylpropanoic acid
A185931-1G
A185931-250MG
A185931-5G
103815-896EA
51
USD
103815-896
103846-470
77084-120
77097-020
77302-002
Boc-DL-β-Aib-OH 98%
Boc-DL-beta-Aib-OH
Precaución: for Research Use Only
Formula:
C₉H₁₇NO₄ Melting Pt: 80…90 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD02094137 CAS Number: 16948-10-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...