3-((tert-Butoxycarbonyl)(methyl)amino)propanoic acid 97%
Supplier: Ambeed
|
Warning
|
A118650-100G
A118650-10G
A118650-250MG
A118650-25G
A118650-1G
A118650-5G
103799-088EA
6
USD
103799-088
77271-370
77271-372
77271-374
77271-376
77311-758
77311-760
3-((tert-Butoxycarbonyl)(methyl)amino)propanoic acid 97%
3-((tert-Butoxycarbonyl)(methyl)amino)propanoic acid
Formula:
C₉H₁₇NO₄ Melting Pt: 59…62 °C Storage Temperature: Ambient |
MDL Number:
MFCD03490508 CAS Number: 124072-61-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...