3-((tert-Butoxycarbonyl)(methoxy)amino)propanoic acid 95%

Supplier: Ambeed
Danger

A204131-1G A204131-5G
103817-526EA 68 USD
103817-526 103848-190 77302-898 77302-900
3-((tert-Butoxycarbonyl)(methoxy)amino)propanoic acid 95%
3-((tert-Butoxycarbonyl)(methoxy)amino)propanoic acid

Precaución: for Research Use Only

Formula: C₉H₁₇NO₅
MW: 219.24 g/mol
Boiling Pt: 327.8 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD04112593
CAS Number: 172299-81-9
UN: 2922
ADR: 8,6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR