2-Cyanoethyl-N,N,N',N'-tetraisopropylphosphordiamidite 97%

Supplier: Ambeed
Danger

Synonyms: Bis(diisopropylamino)(2-cyanoethoxy)phosphine, 3-((Bis(diisopropylamino)phosphino)oxy)propanenitrile

A199325-1G A199325-10G
103817-094EA 9 USD
103817-094 103847-718 77085-764 77550-270
2-Cyanoethyl-N,N,N',N'-tetraisopropylphosphordiamidite 97%
2-Cyanoethyl-N,N,N',N'-tetraisopropylphosphordiamidite

Precaución: for Research Use Only

Formula: C₁₅H₃₂N₃OP
MW: 301.41 g/mol
Boiling Pt: 100 °C (760 mm Hg)
Flash Pt: 60 °C
Storage Temperature: Freezer
MDL Number: MFCD00012213
CAS Number: 102691-36-1
UN: 1993
ADR: 3,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR