3-(2-Ethylhexyloxy)-1,2-propanediol 97%

Supplier: Ambeed
Danger

A709201-25G A709201-500G
103840-690EA 5 USD
103840-690 76844-906 77552-164
3-(2-Ethylhexyloxy)-1,2-propanediol 97%
3-(2-Ethylhexyloxy)-1,2-propanediol

Precaución: for Research Use Only

Formula: C₁₁H₂₄O₃
Boiling Pt: 325.831 °C (760 mm Hg)
Flash Pt: 150.858 °C
Storage Temperature: Ambient
MDL Number: MFCD03790878
CAS Number: 70445-33-9
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR