2-(Trifluoromethyl)nicotinic acid 98%

Supplier: Ambeed
Warning

Synonyms: 2-Trifluoromethylnicotinic acid

A331153-1G A331153-25G A331153-100G A331153-10G
103825-574EA 11 USD
103825-574 103836-538 76839-524 76839-526 77154-700 77154-702
2-(Trifluoromethyl)nicotinic acid 98%
2-(Trifluoromethyl)nicotinic acid

Precaución: for Research Use Only

Formula: C₇H₄F₃NO₂
Melting Pt: 184…188 °C
Flash Pt: 111 °C
Storage Temperature: Ambient
MDL Number: MFCD01862052
CAS Number: 131747-43-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR