2-(Trifluoromethoxy)phenylboronic acid 97%

Supplier: Ambeed
Warning

Synonyms: 2-Trifluoromethoxyphenylboronic acid, 2-(Trifluoromethoxy)benzeneboronic acid, (2-(Trifluoromethoxy)phenyl)metaboric acid, Dihydroxy(2-(trifluoromethoxy)phenyl)borane

A133205-1G A133205-25G A133205-5G A133205-100G A133205-10G
103801-844EA 5 USD
103801-844 77314-470 77314-472 77314-474 77747-952 77747-953
2-(Trifluoromethoxy)phenylboronic acid 97%
2-(Trifluoromethoxy)phenylboronic acid

Precaución: for Research Use Only

Formula: C₇H₆BF₃O₃
MW: 205.93
Boiling Pt: 267.2 °C (760 mm Hg)
Melting Pt: 118…120 °C
Flash Pt: 115.4 °C
Storage Temperature: Ambient
MDL Number: MFCD01320763
CAS Number: 175676-65-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR