Thiophene-2-methylamine 97%
Supplier: Ambeed
|
Danger
|
Synonyms:
2-Thiophenemethylamine, 2-(Aminomethyl)thiophene
A174883-25G
A174883-5G
A174883-100G
A174883-10G
A174883-1G
103806-984EA
5
USD
103806-984
103845-284
77082-594
77082-596
77286-980
77286-982
77286-984
77301-358
Thiophene-2-methylamine 97%
Thiophene-2-methylamine
Precaución: for Research Use Only
Formula:
C₅H₇NS Boiling Pt: 194.5 °C (760 mm Hg) Flash Pt: 74 °C Storage Temperature: Ambient |
MDL Number:
MFCD00005460 CAS Number: 27757-85-3 UN: 2735 ADR: 8,Ⅲ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...