2-Thiopheneethanol 98%

Supplier: Ambeed
Warning

Synonyms: 2-(2-Thienyl)ethanol, Thiophen-2-ethanol

A331878-25G A331878-1000G A331878-100G A331878-10G A331878-500G A331878-5G
103836-558EA 5 USD
103836-558 76839-550 76978-264 76978-266 76978-268 76978-270 77306-426
2-Thiopheneethanol 98%
2-Thiopheneethanol

Precaución: For research use only

Formula: C₆H₈OS
MW: 128.19 g/mol
Boiling Pt: 223 °C (760 mm Hg)
Flash Pt: 101.1 °C
Storage Temperature: Ambient
MDL Number: MFCD00005462
CAS Number: 5402-55-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR