Thiazole-2-carbaldehyde 97%

Supplier: Ambeed
Warning

Synonyms: 2-Thiazolecarboxaldehyde, Thiazole-2-carboxaldehyde

A105381-10G A105381-1G A105381-25G A105381-5G A105381-100G
103796-882EA 9 USD
103796-882 103822-610 77283-354 77283-356 77283-358 77309-482 77747-785
Thiazole-2-carbaldehyde 97%
Thiazole-2-carbaldehyde

Precaución: for Research Use Only

Formula: C₄H₃NOS
MW: 113.13 g/mol
Boiling Pt: 218.3 °C (760 mm Hg)
Flash Pt: 68 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00142924
CAS Number: 10200-59-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR