2-Quinoxalinol 98%

Supplier: Ambeed
Warning

Synonyms: 2-Quinoxalinone, 2(1H)-Quinoxalinone, 2-Hydroxyquinoxaline, quinoxalin-2-ol

A103251-100G A103251-25G A103251-500G A103251-1000G A103251-10G
103807-862EA 107 USD
103807-862 77257-342 77257-344 77257-346 77283-070 77570-192
2-Quinoxalinol 98%
2-Quinoxalinol

Precaución: for Research Use Only

Formula: C₈H₆N₂O
MW: 146.15 g/mol
Melting Pt: 271…272 °C
Flash Pt: 173.6 °C
Storage Temperature: Ambient
MDL Number: MFCD00006722
CAS Number: 1196-57-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR