2-Quinoxalinecarboxylic acid 97%

Supplier: Ambeed
Warning

A131240-10G A131240-500G A131240-1G A131240-25G A131240-5G
103801-550EA 9 USD
103801-550 77258-474 77258-476 77298-468 77314-030 77314-032
2-Quinoxalinecarboxylic acid 97%
2-Quinoxalinecarboxylic acid

Precaución: for Research Use Only

Formula: C₉H₆N₂O₂
MW: 174.16 g/mol
Melting Pt: 208 °C
Flash Pt: 168.6 °C
Storage Temperature: Ambient
MDL Number: MFCD00012334
CAS Number: 879-65-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR