2-Pyridone-6-carboxaldehyde 95%

Supplier: Ambeed
Warning

Synonyms: 6-Oxo-1,6-dihydropyridine-2-carbaldehyde, 1,6-Dihydro-6-oxopyridine-2-carboxaldehyde

A129538-100MG A129538-250MG A129538-1G A129538-5G
103801-300EA 241 USD
103801-300 76782-258 76782-260 77313-684 77313-686
2-Pyridone-6-carboxaldehyde 95%
2-Pyridone-6-carboxaldehyde

Precaución: for Research Use Only

Formula: C₆H₅NO₂
MW: 123.11 g/mol
Boiling Pt: 334.629 °C (760 mm Hg)
Melting Pt: 156…159 °C
Flash Pt: 156.178 °C
Storage Temperature: Refrigerator
MDL Number: MFCD10688270
CAS Number: 358751-77-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR