2-Phenyl-1,3-thiazole-4-carbaldehyde 95%

Supplier: Ambeed
Warning

Synonyms: 2-Phenylthiazole-4-carbaldehyde

A211674-1G A211674-250MG
103818-132EA 33 USD
103818-132 103848-828 77087-250 77275-172
2-Phenyl-1,3-thiazole-4-carbaldehyde 95%
2-Phenyl-1,3-thiazole-4-carbaldehyde

Precaución: for Research Use Only

Formula: C₁₀H₇NOS
MW: 189.24 g/mol
Boiling Pt: 353 °C (760 mm Hg)
Melting Pt: 51 °C
Flash Pt: 167.3 °C
Storage Temperature: Refrigerator
MDL Number: MFCD02681934
CAS Number: 20949-81-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR