2-Phenylpyrrolidine 97%

Supplier: Ambeed
Warning

A166328-1G A166328-5G A166328-10G
103806-052EA 23 USD
103806-052 103844-314 77300-830 77300-832 77567-420
2-Phenylpyrrolidine 97%
2-Phenylpyrrolidine

Precaución: For research use only.

Formula: C₁₀H₁₃N
MW: 147.2200 g/mol
Boiling Pt: 237 °C (760 mm Hg)
Flash Pt: 98.3 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01631835
CAS Number: 1006-64-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR