2-Phenylpyridine 98%

Supplier: Ambeed
Warning

A149115-100G A149115-10G A149115-500G A149115-25G A149115-5G
103842-124EA 6 USD
103842-124 76798-554 76798-556 76798-558 77317-604 77317-606
2-Phenylpyridine 98%
2-Phenylpyridine

Precaución: for Research Use Only

Formula: C₁₁H₉N
MW: 155.2 g/mol
Boiling Pt: 268…270 °C (760 mm Hg)
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00006280
CAS Number: 1008-89-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR