2-Phenylpropionic acid 97%

Supplier: Ambeed
Warning

Synonyms: (±)-2-Phenylpropionic acid, (±)-Hydratropic acid, DL-2-Phenylpropionic acid, (±)-2-Phenylpropanoic acid

A270985-100G A270985-10G A270985-500G A270985-25G A270985-1000G
103852-932EA 5 USD
103852-932 76977-172 76977-174 76977-176 77092-704 77672-398
2-Phenylpropionic acid 97%
2-Phenylpropionic acid

Precaución: for Research Use Only

Formula: C₉H₁₀O₂
MW: 150.18 g/mol
Boiling Pt: 261 °C (760 mm Hg)
Melting Pt: 5 °C
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00002650
CAS Number: 492-37-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR