2-Phenylpropan-2-yl Benzodithioate 97%

Supplier: Ambeed
Danger

Synonyms: 2-Phenylpropan-2-yl benzodithioate

A572602-1G A572602-5G A572602-250MG
103828-810EA 118 USD
103828-810 103839-952 76844-078 76844-080 77254-074
2-Phenylpropan-2-yl Benzodithioate 97%
2-Phenylpropan-2-yl Benzodithioate

Precaución: for Research Use Only

Formula: C₁₆H₁₆S₂
MW: 272.42 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD18252676
CAS Number: 201611-77-0
UN: 3077
ADR: 9 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR