Cumylamine 97% (by GC)

Supplier: Ambeed
Danger

Synonyms: alpha,alpha-Dimethylbenzylamine, Phenylisopropylamine, alpha-Aminoisopropylbenzene, 2-Phenylpropan-2-amine

A161043-10G A161043-1G A161043-25G
103805-444EA 5 USD
103805-444 103843-682 76689-830 77319-646 77319-648
Cumylamine 97% (by GC)
Cumylamine

Precaución: for Research Use Only

Formula: C₉H₁₃N
MW: 135.21 g/mol
Boiling Pt: 197 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD00134680
CAS Number: 585-32-0
UN: 2735
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR