2-(2-Phenylethynyl)aniline 95%

Supplier: Ambeed
Warning

Synonyms: 2-Phenylethynyl-phenylamine

A101776-100MG A101776-250MG A101776-1G A101776-5G
103796-130EA 60 USD
103796-130 103821-818 77093-526 77093-528 77282-830 77282-832
2-(2-Phenylethynyl)aniline 95%
2-(2-Phenylethynyl)aniline

Precaución: for Research Use Only

Formula: C₁₄H₁₁N
MW: 193.244 g/mol
Boiling Pt: 359.7 °C (760 mm Hg)
Melting Pt: 89 °C
Flash Pt: 186 °C
Storage Temperature: Ambient
MDL Number: MFCD00168849
CAS Number: 13141-38-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR