2-Phenylcyclopropanecarboxylic acid 97%

Supplier: Ambeed
Warning

A226797-250MG A226797-100MG
103819-272EA 31 USD
103819-272 103850-180 76874-158 77883-590
2-Phenylcyclopropanecarboxylic acid 97%
2-Phenylcyclopropanecarboxylic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₀O₂
Boiling Pt: 317.1±31.0 °C (760 mm Hg)
Melting Pt: 88…92 °C
Flash Pt: 144.3 °C
Storage Temperature: Ambient
MDL Number: MFCD00066860
CAS Number: 5685-38-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR