Biphenyl-2-carboxaldehyde 97%

Supplier: Ambeed
Warning

Synonyms: 2-Phenylbenzaldehyde, 2-Biphenylcarboxaldehyde

A201849-100G A201849-10G A201849-1G A201849-25G A201849-5G
103817-334EA 17 USD
103817-334 103847-974 76721-478 76721-480 77086-060 77086-062 77086-064
Biphenyl-2-carboxaldehyde 97%
Biphenyl-2-carboxaldehyde

Precaución: for Research Use Only

Formula: C₁₃H₁₀O
MW: 182.218 g/mol
Boiling Pt: 91…92 °C at 0.15 mmHg
Flash Pt: 101.7 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01740431
CAS Number: 1203-68-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR